Learn More
α,α-Diphenyl-4-piperidinomethanol, 98+%, Thermo Scientific™
24.40 € - 74.40 €
Chemical Identifiers
CAS | 115-46-8 |
---|---|
Molecular Formula | C18H21NO |
Molecular Weight (g/mol) | 267.37 |
MDL Number | MFCD00066980 |
InChI Key | ZMISODWVFHHWNR-UHFFFAOYSA-N |
Synonym | azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol |
PubChem CID | 15723 |
IUPAC Name | diphenyl(piperidin-4-yl)methanol |
SMILES | C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O |
Description
Chemical Identifiers
115-46-8 | |
267.37 | |
ZMISODWVFHHWNR-UHFFFAOYSA-N | |
15723 | |
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O |
C18H21NO | |
MFCD00066980 | |
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
diphenyl(piperidin-4-yl)methanol |
Specifications
115-46-8 | |
100.0 | |
White to Beige | |
Authentic | |
C18H21NO | |
13, 895 | |
ZMISODWVFHHWNR-UHFFFAOYSA-N | |
diphenyl(piperidin-4-yl)methanol | |
15723 | |
98+% | |
α, α-Diphenyl-4-piperidinomethanol, 98+% |
98.0 | |
160.0°C to 163.0°C | |
1% max. | |
98% min. (HPLC) | |
MFCD00066980 | |
azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O | |
267.37 | |
267.37 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-092-5
missing translation for 'rtecsNumber' : TN0470000
missing translation for 'tsca' : TSCA
- a,a-Diphenyl-4-piperidinomethanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.