missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl carbonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.
Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.
Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 76°C to 81°C |
Density | 1.3 |
Boiling Point | 301°C to 302°C |
Flash Point | 168°C (334°F) |
Odor | Mild |
Quantity | 250 g |
Beilstein | 1074863 |
Merck Index | 14,3602 |
Solubility Information | Soluble in ethanol,diethyl ether,carbon tetrachloride,acetic acid. Insoluble in water. |
Formula Weight | 214.22 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction