missing translation for 'onlineSavingsMsg'
Learn More
Diphenyl chlorophosphate, 98%, Thermo Scientific Chemicals Product Code.: 10206873

Diphenyl chlorophosphate, 98%, Thermo Scientific Chemicals

Product Code. 10206873
500 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10206873

Brand: Thermo Scientific Acros 117245000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.63
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Color Colorless to Yellow
Density 1.2900g/mL
Boiling Point 314°C to 316°C (272.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5O)2P(O)Cl
Refractive Index 1.5485 to 1.5505
Quantity 500 mL
Beilstein 03, 179
Fieser 01,345; 02,180; 03,133; 11,223
Merck Index 15, 3367
Specific Gravity 1.29
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in most common organic solvents
Viscosity 14 mPa.s (25°C)
Formula Weight 268.63
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diphenyl chlorophosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0459635 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number117245000,117241000,117240250
Lot NumberA0374309 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number117245000,117241000,117240250
Lot NumberA0384105 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number117245000,117241000,117240250
Lot NumberA0418284 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number117245000,117241000,117240250
Lot NumberA0443288 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number117245000,117241000,117240250
compliance-icons
Product Identifier
  • Diphenyl chlorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenyl chlorophosphate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.