missing translation for 'onlineSavingsMsg'
Learn More
Diphenyl phosphorochloridate, 97%, Thermo Scientific Chemicals Product Code.: 11400034

Diphenyl phosphorochloridate, 97%, Thermo Scientific Chemicals

Product Code. 11400034
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11400034

Brand: Thermo Scientific Alfa Aesar A13546.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5',5'-triphosphate, guanosine-cytidine-5',5'-triphosphate, guanosine-uridine-5',5'-triphosphate, diguanosine-5',5'-diphosphate and diguanosine-5',5'-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl phosphorochloridate is used to prepare cyclohexyl-amidophosphoric acid diphenyl ester by reacting with cyclohexylamine. It is also used in the synthesis of alfa-substituted beta-lactams, anhydrides, esters and thioesters. It is also employed in the conversion of aldoximes to nitriles. Further, it is used in the preparation and reaction of enol phosphates, guanosine-adenosine-5′,5′-triphosphate, guanosine-cytidine-5′,5′-triphosphate, guanosine-uridine-5′,5′-triphosphate, diguanosine-5′,5′-diphosphate and diguanosine-5′,5′-triphosphate. In addition to this, it serves as a phosphorylating agent for the preparation of 2-deoxy-D-galactose-3 and -6 phosphoric acids.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.633
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Density 1.296
Boiling Point 145°C to 147°C (1 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.55
Quantity 50 g
UN Number UN3265
Beilstein 654130
Sensitivity Moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 268.64
Percent Purity 97%
Chemical Name or Material Diphenyl phosphorochloridate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Diphenyl phosphorochloridate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.