missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl sulfide, 98%, Thermo Scientific Chemicals

Product Code. 11429003
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl sulfide is used as a raw material for triaryl sulfonium photoinitiator, intermediates for agrichemicals, catalyst, metal extractants.

Solubility
Soluble in ether, benzene, carbon disulphide.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-66-2
Molecular Formula C12H10S
Molecular Weight (g/mol) 186.27
MDL Number MFCD00003064
InChI Key LTYMSROWYAPPGB-UHFFFAOYSA-N
Synonym diphenyl sulfide, phenyl sulfide, phenylthiobenzene, diphenyl sulphide, diphenyl thioether, benzene, 1,1'-thiobis, diphenylmercaptan, diphenylthiamethane, diphenyl monosulfide, sulfide, diphenyl
PubChem CID 8766
ChEBI CHEBI:38959
IUPAC Name phenylsulfanylbenzene
SMILES S(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -40°C
Density 1.118
Boiling Point 295°C to 296°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.6325
Quantity 100 g
Beilstein 1907932
Merck Index 14,7314
Solubility Information Soluble in ether,benzene,carbon disulphide.
Formula Weight 186.28
Percent Purity 98%
Chemical Name or Material Diphenyl sulfide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.