missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as a high temperature solvent. Such high temperature solvents are useful for processing highly rigid polymers, e.g., PEEK, which only dissolve in very hot solvents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 123°C to 130°C |
Density | 1.36 |
Boiling Point | 377°C to 379°C |
Flash Point | 184°C (363°F) |
Odor | Odorless |
Quantity | 250 g |
Beilstein | 1910573 |
Merck Index | 14,3238 |
Solubility Information | Insoluble in water. |
Formula Weight | 218.27 |
Show More |
Safety and Handling


Product Identifier
- Diphenyl sulfone
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Diphenylsulfone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Diphenyl sulfone, 99+%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction