Learn More
Diphenylcyclopropenone, 98%, Thermo Scientific Chemicals
60.60 € - 883.40 €
Chemical Identifiers
CAS | 886-38-4 |
---|---|
Molecular Formula | C15H10O |
Molecular Weight (g/mol) | 206.24 |
MDL Number | MFCD00001311 |
InChI Key | HCIBTBXNLVOFER-UHFFFAOYSA-N |
Synonym | diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec |
PubChem CID | 65057 |
ChEBI | CHEBI:53074 |
IUPAC Name | 2,3-diphenylcycloprop-2-en-1-one |
SMILES | O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10256230
|
Thermo Scientific Acros
175820010 |
1 g | Glass bottle |
60.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10317740
|
Thermo Scientific Acros
175820050 |
5 g | Glass bottle |
198.99 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954851
|
Thermo Scientific Acros
175820250 |
25 g | Glass bottle |
883.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
886-38-4 | |
206.24 | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
65057 | |
2,3-diphenylcycloprop-2-en-1-one |
C15H10O | |
MFCD00001311 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
CHEBI:53074 | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
886-38-4 | |
Beige to Orange-Pink | |
97.5% min. (HPLC) | |
MFCD00001311 | |
15,3338 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol, ethanol, acetone, chloroform,, dichloromethane, cyclohexane, benzene, hexane,, petroleum ether | |
O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
206.24 | |
CHEBI:53074 | |
98% | |
Diphenylcyclopropenone |
117.0°C to 121.0°C | |
Authentic | |
C15H10O | |
07,IV,1736 | |
diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec | |
HCIBTBXNLVOFER-UHFFFAOYSA-N | |
2,3-diphenylcycloprop-2-en-1-one | |
65057 | |
206.24 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-948-4
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.