missing translation for 'onlineSavingsMsg'
Learn More

Diphenyleneiodonium chloride, Tocris Bioscience™

Product Code. 18084912
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GPR3 agonist; also inhibits NOS and NADPH oxidases

GPR3 agonist (EC50 = 1 μM); activates adenylate cyclase through GPR3 but not GPR6 or GPR12. Also induces Ca2+ mobilization and β-arrestin receptor internalization. Binds strongly to flavoproteins; inhibits several enzymes, including NO synthase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Also inhibits platelet aggregation.

Chemical Identifiers

CAS 4673-26-1
Molecular Formula C12H8ClI
Molecular Weight (g/mol) 314.55
InChI Key FCFZKAVCDNTYID-UHFFFAOYSA-M
Synonym diphenyleneiodonium chloride, dibenziodolium chloride, unii-7m9d81yz2n, dpi, 8? 3-iodatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2,4,6,9,11-hexaen-8-ylium chloride, dibenziodolium, chloride, d0n0gj, c12h8i.cl, dibenzo b,d iodolium chloride, 2,2'-biphenylyleneiodonium chloride
PubChem CID 2733504
ChEBI CHEBI:77967
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[I+]2.[Cl-]

Specifications

Quantity 50 mg
Formula Weight 314.55
Percent Purity >99%
Chemical Name or Material Diphenyleneiodonium chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'