missing translation for 'onlineSavingsMsg'
Learn More

Diphenylgermanium dichloride, 98+%, Thermo Scientific Chemicals

Product Code. 11327788
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diphenylgermanium dichloride is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1613-66-7
Molecular Formula C12H14Cl2Ge
Molecular Weight (g/mol) 301.77
MDL Number MFCD00013587
InChI Key YROQISGFJISKEN-UHFFFAOYSA-N
Synonym diphenyldichlorogermane, diphenylgermanium dichloride, germane, dichlorodiphenyl, dichlorodiphenylgermene, dichloro diphenyl germane, acmc-1bssq, germane,dichlorodiphenyl, germanium diphenyl dichloride
PubChem CID 74167
IUPAC Name dichloro(diphenyl)germane
SMILES Cl.Cl.[GeH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 9°C
Density 1.415 g/mL
Boiling Point 107°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 297.73
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Diphenylgermanium dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'