missing translation for 'onlineSavingsMsg'
Learn More

Diphenylsilane, 97%, Thermo Scientific Chemicals

47.94 € - 444.21 €

Chemical Identifiers

CAS 775-12-2
Molecular Formula C12H10Si
Molecular Weight (g/mol) 182.297
MDL Number MFCD00003002
InChI Key BPYFPNZHLXDIGA-UHFFFAOYSA-N
Synonym diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
PubChem CID 6327659
IUPAC Name cyclohexa-2,5-dien-1-ylidene(phenyl)silanide
SMILES C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404300
View Documents
Thermo Scientific Alfa Aesar
A10884.06
5 g
47.94 €
5g
Please to purchase this item. Need a web account? Register with us today!
11414300
View Documents
Thermo Scientific Alfa Aesar
A10884.14
25 g
137.69 €
25g
Please to purchase this item. Need a web account? Register with us today!
11424300
View Documents
Thermo Scientific Alfa Aesar
A10884.22
100 g
444.21 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thiocarbonyl derivatives of secondary alcohols are readily reduced by diphenylsilane in a radical chain process at room temperature using triethylborane-air as an initiator. An improved radical chain procedure for the deoxygenation of secondary and primary alcohols using diphenylsilane as hydrogen atom donor and triethylborane-air as initiator. Diphenylsilane is a reagent in the invention of radical reactions for deoxygenation of alcohols via their thiocarbonyl derivatives, deamination via isonitriles, and dehalogenation of bromo- and iodo- compounds by radical chain chemistry. Fluorescent film sensor for vapor-phase nitroaromatic explosives via monolayer assembly of oligo(diphenylsilane) on glass plate surfaces. Reductions of carboxylic acid derivatives by silanes in the presence of rhodium complexes were studied. Carboxylic esters were reduced to alcohols by diphenylsilane catalyzed by [RhCl (cod)] 2/4PPh 3 or [RhCl (PPh 3) 3] at room temperature in up to 99% yields. Sequential C- Si bond formations from diphenylsilane and its application to silanediol peptide isostere precursors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiocarbonyl derivatives of secondary alcohols are readily reduced by diphenylsilane in a radical chain process at room temperature using triethylborane-air as an initiator. An improved radical chain procedure for the deoxygenation of secondary and primary alcohols using diphenylsilane as hydrogen atom donor and triethylborane-air as initiator. Diphenylsilane is a reagent in the invention of radical reactions for deoxygenation of alcohols via their thiocarbonyl derivatives, deamination via isonitriles, and dehalogenation of bromo- and iodo- compounds by radical chain chemistry. Fluorescent film sensor for vapor-phase nitroaromatic explosives via monolayer assembly of oligo(diphenylsilane) on glass plate surfaces. Reductions of carboxylic acid derivatives by silanes in the presence of rhodium complexes were studied. Carboxylic esters were reduced to alcohols by diphenylsilane catalyzed by [RhCl (cod)] 2/4PPh 3 or [RhCl (PPh 3) 3] at room temperature in up to 99% yields. Sequential C- Si bond formations from diphenylsilane and its application to silanediol peptide isostere precursors.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, haevy metal powders.
Specifications

Chemical Identifiers

775-12-2
182.297
BPYFPNZHLXDIGA-UHFFFAOYSA-N
6327659
C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2
C12H10Si
MFCD00003002
diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
cyclohexa-2,5-dien-1-ylidene(phenyl)silanide

Specifications

95°C to 97°C (13mmHg)
C12H10Si
MFCD00003002
98°C (208°F)
Decomposes in water.
C1=CC=C(C=C1)[Si-]=C2C=C[CH+]C=C2
182.297
184.32
1.579
Diphenylsilane
775-12-2
(C6H5)2SiH2
2935887
diphenylsilane, silane, diphenyl, diphenylsilicon, benzene, 1,1'-silylenebis, diphenyl silane, di phenyl silicon, diphenylsilylene radical, ph 2sih2
BPYFPNZHLXDIGA-UHFFFAOYSA-N
cyclohexa-2,5-dien-1-ylidene(phenyl)silanide
6327659
97%
0.996
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 212-271-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.