missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenylsilanediol, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Descripción
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
Solubility
Reacts with water.
Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.
Solubility
Reacts with water.
Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
Especificaciones
Especificaciones
Melting Point | ∼150°C (decomposition) |
Quantity | 25 g |
Linear Formula | (C6H5Si(OH)2 |
UN Number | UN1325 |
Beilstein | 2523445 |
Formula Weight | 216.31 |
Percent Purity | 98% |
Odor | Odorless |
Chemical Name or Material | Diphenylsilanediol |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido