missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin dichloride, 90+%, Thermo Scientific Chemicals

Product Code. 11334829
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in the manufacture of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-99-5
Molecular Formula C12H12Cl2Sn
Molecular Weight (g/mol) 345.84
MDL Number MFCD00000516
InChI Key WSXPROXKZIMZIU-UHFFFAOYSA-N
Synonym diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr
PubChem CID 14342
IUPAC Name dichloro(diphenyl)stannane
SMILES Cl.Cl.[SnH2](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 41°C to 43°C
Color White
Boiling Point 333°C to 337°C (decomposition)
Flash Point >150°C (302°F)
Linear Formula (C6H5)2SnCl2
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Sparingly Soluble (2.4E-3g/L) (25°C),Calc.
Formula Weight 343.81
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Diphenyltin dichloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'