missing translation for 'onlineSavingsMsg'
Learn More

Diphenyltin oxide, Thermo Scientific Chemicals

Product Code. 11314557
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2273-51-0
Molecular Formula C12H10OSn
Molecular Weight (g/mol) 288.92
MDL Number MFCD00058851
InChI Key VPPWQRIBARKZNY-UHFFFAOYSA-N
Synonym diphenyltin oxide, diphenyltin oxide polymer, stannane, diphenyloxo-, polymer, stannane, oxodiphenyl-, polymer, diphenylstannanone, oxodiphenylstannane, oxo diphenyl stannane, oxo diphenyl tin, stannane, polymer, iv
PubChem CID 75280
IUPAC Name oxo(diphenyl)tin
SMILES O=[Sn](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 320°C (decomposition)
Quantity 5 g
UN Number UN3146
Formula Weight 288.9
Physical Form Powder
Chemical Name or Material Diphenyltin oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'