missing translation for 'onlineSavingsMsg'
Learn More

Diphenylzinc, 98+%, Thermo Scientific Chemicals

Product Code. 11319129
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Diphenylzinc is used as the synthetic equivalent of ph- synthon. It is also useful in a Michael 1,4-addition reactions. It is used as a catalyst for propylene oxide polymerization in benzene medium. Further, it is a reactive coupling compound in Negishi cross-coupling reactions. In addition to this, it utilized in the preparation of arylcopper, arylsilver and arylgold compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078-58-6
Molecular Formula C12H10Zn
Molecular Weight (g/mol) 219.59
MDL Number MFCD00014071
InChI Key BUNROVZNGITPIX-UHFFFAOYSA-N
Synonym diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2
PubChem CID 517896
IUPAC Name zinc;benzene
SMILES [Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1

Specifications

Melting Point 107°C
Boiling Point 280°C to 285°C
Quantity 1 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Solubility Information Soluble in most organic solvents.
Formula Weight 219.58
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Diphenylzinc

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.