missing translation for 'onlineSavingsMsg'
Learn More

Dipyridamole, Tocris Bioscience™

Product Code. 18034302
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Adenosine transport inhibitor

Coronary vasodilator; adenosine transport inhibitor. Phosphodiesterase inhibitor (IC50 values are 0.37, 0.38, 0.45, 0.9 and 4.5 μM for PDE11, 6, 10, 5 and 8 respectively). Inhibits ENT1 and ENT2 (IC50 = 144.8 nM and Ki = 8.18 nM for ENT1).

Specifications

CAS 58-32-2
Molecular Formula C24H40N8O4
Quantity 500 mg
Synonym dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil
InChI Key IZEKFCXSFNUWAM-UHFFFAOYSA-N
SMILES C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO
IUPAC Name 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
Molecular Weight (g/mol) 504.636
PubChem CID 3108
ChEBI CHEBI:4653
Formula Weight 504.63
Percent Purity >99%
Chemical Name or Material Dipyridamole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'