missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Directional Arrow Tape

Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.

Brand:  Brady™ 275100

52.94 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 12727946

  • 62.15 € / 1 roll
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Directional Arrow Tape
Black on Yellow
Tape
Pipe Marker
1300 Markers/Roll
-40°F to +230°F
0.0039 in.
33 m x 25 mm
33 m
25 mm
ARROWTAPE:BLACK/YELLOW25MMX33M
Permanent Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Pipe and valve marking
1300 Pk.
-40°C to +110°C
0.099 mm
108.27 ft.
0.98 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.