Learn More
Brady™ Directional Arrow Tape
Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Brand:  Brady™ 275109
 View more versions of this product
Product Code. 12767916
  • 71.00 € / 1 roll
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Directional flow arrow tape can enhance safety, improve operating effciency and provides 360 degree visibility.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379
Directional Arrow Tape
White on Black
Tape
Pipe Marker
1300 Markers/Roll
-40°F to +230°F
0.0039 in.
33 m x 25 mm
33 m
25 mm
ARROWTAPE:BK/WHITEARROW-25MMX33M
Permanent Acrylic, Permanent, Pressure-sensitive
Gloss
Polyester
Pipe and valve marking
1300 Pk.
-40°C to +110°C
0.099 mm
108.27 ft.
0.98 in.