Learn More
VELP Scientifica Disc 1 for Capsules for Autosampler
30-position disc for capsules for liquids or semi-solid samples (expandable with an additional disc A00000420 - up to 59 samples).
Brand:  VELP Scientifica A00000420
Product Code. 30003202
  • 728.00 € / Each
Explore more special offers
Description
Description
  • Additional 30-position disc for capsules, for liquids or semi-solid samples
  • Suitable for the CN 802 Carbon/Nitrogen Analyser
  • The VELP C/N boasts exceptional efficiency, enabling the highest sample throughput. This is made possible by theelectronic autosampler, capable of processing up to 117 samples at a time.
Specifications
Specifications
Autosampler Disc