Learn More
VELP Scientifica Disc 2 for Capsules for Autosampler
30-position disc for capsules for liquids or semi-solid samples (to be used in combination with disc A00000390).
Brand:  VELP Scientifica A00000390
Product Code. 30003201
  • 677.00 € / Each
Explore more special offers
Description
Description
  • Additional 30-position disc for capsules, for liquids or semi-solid samples
  • Suitable for the CN 802 Carbon/Nitrogen Analyser
  • The VELP C/N boasts exceptional efficiency, enabling the highest sample throughput. This is made possible by theelectronic autosampler, capable of processing up to 117 samples at a time.
Specifications
Specifications
Autosampler Disc