missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Stuart™ Discharge Tube for Pressmatic Bottle Top Dispenser
Choose a discharge tube for Model PD10P or PD60P Pressmatic Bottle Top Dispenser
50.10 € - 63.30 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12437772
|
Cole-Parmer™
99955-60 |
Stuart PD10P Pressmatic |
50.10 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
12447772
|
Cole-Parmer™
16223-99 |
Stuart PD60P Pressmatic | N/A | ||||||
Description
- For use with a Stuart™ Model PD10P or PD60P Pressmatic Bottle Top Dispensers
- PD10P works with smaller volumes from 2 to 10mL
- PD60P works with volumes between 10 and 60mL
- Compatible with dilute acids, buffers, polar solvents, biological solutions
Specifications
Discharge Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title