Learn More
Disinfectant Wipe
Brand:  Medimark Scientific XTM353
Product Code. 15116686
  • 22.45 € / Pack of 100
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Lab Disinfectant Wipe
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Chemgene HLD4 Wipes
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : QUATERNARY AMMONIUMCOMPOUNDS