missing translation for 'onlineSavingsMsg'
Learn More

Disodium hydrogen phosphate dodecahydrate, BAKER ANALYZED™, J.T. Baker™

Product Code. 10793811
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point 35°C
CAS 10039-32-4
pH 8.7 to 9.3 (5% Solution at 25°C)
Quantity 1 kg
Assay Percent Range 98.0 to 101.0%
Molecular Formula H25Na2O16P
MDL Number MFCD00149181
InChI Key DGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium dodecahydrate hydrogen phosphate
Molecular Weight (g/mol) 358.14
Density 1.52 g/cm³ (20°C)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'