Learn More
BRAND™ Dispenser Box for Schulke Wipes
Dispenser Box designed to hold disinfecting fleece wipes.
Brand:  BRAND™ 44875
21.84 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15431945
  • 27.70 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For high-volume use, wipes can be soaked with liquid disinfectant directly in the handy dispenser box
  • Lasts up to 28 days
  • The usage of the surface disinfecting detergent Pursept™ A Xpress are recommended
  • Can hold up to 70 wipes
  • Refillable and firmly closable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dispenser Box
230 x 280 mm
Holds 70 fleece wipes
Empty
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Dispenser Box for Schulke Wipes > Dimensions (L x W): 230 x 280 mm