Learn More
Kartell™ Dispenser Dosipet HF
Dosing devices are designed for specific use with hydrofluoric acid solutions
Brand:  Kartell™ 8308100
Product Code. 10151813
  • 1860.00 € / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The internal components that come into contact with these solutions are made with high quality and high resistance materials (PTFE and platinum-iridium), completely inert with hydrofluoric acid
  • Complete with PTFE piston
  • CE marked
  • IVD
  • Specifications
    Specifications
    2.5 to 25 mL
    Dosipet HF-25
    45 GL
    0.5 mL
    ± 0.15
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    Special Offers
    Special Offers
    Product Content Correction
    Product Title
    Kartell™ Dispenser Dosipet HF > Model: Dosipet HF-25