missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Dispenser tips

Robust, stable, and durable dispenser tips for several brands of pipettors. For dispensing of fluids with high viscosity and high vapor pressure.

Brand:  Fisherbrand™ 702609

100.96 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 15163646

  • 125.00 € / Pack of 100
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • These disposable tips are suitable for the following multiple dispensers:
  • BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch/ S
  • Eppendorf Multipette™ 4780
  • Gilson Distriman™ and Gilson Repetman™
  • Ritter Ripette™ pro
  • EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
  • STARLAB RepeatOne™
TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.1 mL
ISO 8655
Dispenser Tip
Eppendorf Multipette 4780, Other Compatible System
Non-sterile
For dispensing of fluids with high viscosity, high vapor pressure
Single-use
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Dispenser tips > Capacity: 0.1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.