Learn More
Masterflex™ Straight Tubing Connector
With barbed fittings.
Brand:  Masterflex™ 40622-57
 View more versions of this product
Product Code. 15531295
  • 26.90 € / Pack of 10
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from polypropylene
  • Ethylene Oxide/Gamma sterilized
Specifications
Specifications
Straight Tubing Connector
0.48 cm
ETO or Gamma
0.19 in.
Polypropylene
Barbed fittings