missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispensette™ S Digital Bottletop Dispensers
GREENER_CHOICE

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

518.00 € - 805.00 €

Specifications

Product Type Dispenser
Autoclavable Autoclavable (121°C)
Certifications/Compliance Certifications: ISO 9001
Color Red
Display Digital
View More Specs

Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual

Products 12
Product Code Brand Description Volume (Metric) Increments Accuracy Price Quantity & Availability  
Product Code Brand Description Volume (Metric) Increments Accuracy Price Quantity & Availability  
15408083
GREENER_CHOICE
View Documents
BRAND™
4600361
With recirculation valve 5 to 50 mL 0.2 mL 0.5% (250μL)
805.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15488073
GREENER_CHOICE
View Documents
BRAND™
4600351
With recirculation valve 2.5 to 25 mL 0.1 mL 0.5% (125μL)
718.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
15478063
GREENER_CHOICE
View Documents
BRAND™
4600310
Without recirculation valve 0.1 to 1 mL 0.005 mL 0.6% (6μL)
518.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
15468073
GREENER_CHOICE
View Documents
BRAND™
4600341
With recirculation valve 1 to 10 mL 0.05 mL 0.5% (50μL)
569.00 €
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
15418073
GREENER_CHOICE
View Documents
BRAND™
4600320
Without recirculation valve 0.2 to 2 mL 0.01 mL 0.5% (10μL)
518.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
15448073
GREENER_CHOICE
View Documents
BRAND™
4600331
With recirculation valve 0.5 to 5 mL 0.02 mL 0.5% (25μL)
585.00 €
Each
In Stock
to see stock.
Add to basket
 
15408073
GREENER_CHOICE
View Documents
BRAND™
4600311
With recirculation valve 0.1 to 1 mL 0.005 mL 0.6% (6μL)
580.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
15428073
GREENER_CHOICE
View Documents
BRAND™
4600321
With recirculation valve 0.2 to 2 mL 0.01 mL 0.5% (10μL)
580.00 €
Each
In Stock
to see stock.
Add to basket
 
15438073
GREENER_CHOICE
View Documents
BRAND™
4600330
Without recirculation valve 0.5 to 5 mL 0.02 mL 0.5% (25μL)
518.00 €
Each
In Stock
to see stock.
Add to basket
 
15458073
GREENER_CHOICE
View Documents
BRAND™
4600340
Without recirculation valve 1 to 10 mL 0.05 mL 0.5% (50μL)
518.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
15478073
GREENER_CHOICE
View Documents
BRAND™
4600350
Without recirculation valve 2.5 to 25 mL 0.1 mL 0.5% (125μL)
656.00 €
Each
In Stock
to see stock.
Add to basket
 
15498073
GREENER_CHOICE
View Documents
BRAND™
4600360
Without recirculation valve 5 to 50 mL 0.2 mL 0.5% (250μL)
697.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
Description

Description

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for g

  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Easy Calibration technique for calibration adjustment in seconds without tools
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking

Warranty

One year

Specifications

Specifications

Dispenser
Certifications: ISO 9001
Digital
Autoclavable (121°C)
Red
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
Videos
SDS
Documents

Documents

Product Certifications