Learn More
BRAND™ Dispensing Cylinder with Valve Block
For use with Titrette™ Bottletop Burette. Accommodates all serial number ranges.
126.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15693987
|
BRAND™
707533 |
10 mL Titrette bottletop burette |
126.00 €
Each |
In Stock
Log in to see stock. |
|||||
11663219
|
BRAND™
707537 |
50 mL Titrette bottletop burette |
126.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- For use with Titrette™ Bottletop Burette
- Fits all serial number ranges
- *For 50mL instruments with serial numbers beginning with 01-12 AND letters A-J, must order both Dispensing cylinder and Titrating tube as a pair.
Specifications
Dispensing Cylinder | |
Valve head for titrette |