missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Dispensing System and Dip Tube

Dispensing system and dip tube

362.25 € - 385.00 €

Specifications

Product Type Returnable Container Accessory
Quantity 1
View More Specs

Products 3
Product Code Brand Quantity Capacity (Metric) For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Quantity Capacity (Metric) For Use With (Equipment) Price Quantity & Availability  
16871210
View Documents
Honeywell™
89857H-1EA
1 18 L For 18 L Drum without Dip Tube
385.00 €
1 pack
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
16881210
View Documents
Honeywell™
89864H-1EA
1 45 L For 45 L Drum without Dip Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16891210
View Documents
Honeywell™
89889H-1EA
1 200 L For 200 L Drum without Dip Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

Returnable Container Accessory
1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.