missing translation for 'onlineSavingsMsg'
Learn More

CellPath Dispocut™ Disposable Cut-Up Boards

For routine and high risk specimens

Brand:  CellPath CGB-0402-52A

Product Code. 12816653

  • 218.00 € / Pack of 24
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Disposable by incineration
  • Manufactured from 3mm, three ply foam filled board for strength and rigidity
  • High degree of moisture resistance
Specifications

Specifications

Dispocut Board White
Single-use
297 x 210 mm
White
Routine and high risk specimens
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CellPath Dispocut™ Disposable Cut-Up Boards > Dimensions: 297L x 210mmW

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.