Learn More
Epredia™ Disposable Blade Carriers
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Brand: Epredia™ 705800
1763.75 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
Disposable Blade Carriers- Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
- Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
- Quick clamp system for high- and low-profile blades
- Adjustable clearance angle
- Moveable knife guard covers the blade entirely in any position
- Knife carrier base included
Specifications
Blade carrier E | |
Blade Carrier E | |
HM 310, HM 315 and HM 325 |