Learn More
Epredia™ Disposable Blade Carriers
Product Code. 12647876 Shop All Epredia Products
Click to view available options
Product Type:
Blade Carrier E
Disposable Blade Carrier E
For Use With (Equipment):
Dovetail Style Unit
HM 310, HM 315 and HM 325
Unit Size:
Each
Product Code. 12647876
Brand: Epredia™ 705050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Utilize low- and high-profile blades with the quick clamp system of Epredia™ Disposable Blade Carriers.
Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
  • Quick clamp system for high- and low-profile blades
  • Adjustable clearance angle
  • Moveable knife guard covers the blade entirely in any position
  • Knife carrier base included
TRUSTED_SUSTAINABILITY
Specifications
Description E Type blade carrier for dovetail style units
Product Type Disposable Blade Carrier E
For Use With (Equipment) Dovetail Style Unit