missing translation for 'onlineSavingsMsg'
Learn More
Kolmi Hopen™ Disposable Healthcare Tunic with Jersey Collar and Cuffs Product Code.: 17720179
Estimated Shipment: 11-06-2025
to see stock.
Add to basket

Kolmi Hopen™ Disposable Healthcare Tunic with Jersey Collar and Cuffs

Product Code. 17720179
Healthcare Tunic with Jersey Collar and Cuffs; Closure Type: Press Studs; Color: Navy Blue; Disposable: Yes; Material: Polypropylene; Size: Large
Click to view available options
Healthcare Tunic with Jersey Collar and Cuffs; Closure Type: Press Studs; Color: Navy Blue; Disposable: Yes; Material: Polypropylene; Size: Large
This item is not returnable. View return policy

Product Code. 17720179

Brand: Kolmi Hopen™ J40895/L

Estimated Shipment: 11-06-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Disposable tunic with two pockets, providing extra protection for healthcare workers.

  • Light and breathable polypropylene material
  • Loose shape allows user to maintain good movement
  • Optimally covers upper body
  • Jersey collar and cuffs
  • Pockets for convenient carrying
  • Individual packaging for hygiene and controlled distribution
  • 40 g/m2 fabric weight

Specifications

Size Large
Material Polypropylene
Color Navy
Closure Type Press Studs
Disposable Single-use
No. of Pockets 2
Sleeve Style Long Sleeves
For Use With (Application) Dental, Medical
No. per Pack 50 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kolmi Hopen™ Disposable Healthcare Tunic with Jersey Collar and Cuffs >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.