Learn More
Fisherbrand™ Disposable Inoculating Loops
Product Code. 17793667 Shop All Fisherbrand Products
Click to view available options
Volume:
1 μL
10 μL
Sterility:
Sterile
Quantity:
Cs of 125 (5 Pks. of 25/ Individually Packed)
Cs of 250 (5 Pks. of 50/ Individually Packed)
Unit Size:
Pack of 125
Pack of 250
Product Code. 17793667
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
  • Ideal for anaerobic microbiology applications where deep agar inoculation is required
  • Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
  • High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
  • Rigid enough to penetrate the media for subsurface inoculation
  • Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
  • Can be used under hoods without danger
  • Color-coded for ease of size identification
Specifications
Disposable Single-use
Volume 10 μL
Material Polystyrene
Sterility Sterile
Quantity Cs of 125 (5 Pks. of 25/ Individually Packed)
Autoclavable Not Autoclavable