Learn More
Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KBr window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
Brand: Thermo Scientific Chemicals 043937.KM
252.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Size: 5 x 10cm
- KBr window
- 9.5mm aperture
Specifications
Disposable IR Cards | |
25 Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only