missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KCl window, 19mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

Brand:  Thermo Scientific Chemicals 043932.KM

196.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15476565

  • 196.98 € / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Size: 5 x 10cm
  • KCl window
  • 19mm aperture
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Disposable IR Cards
25 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KCl window, 19mm aperture > 25pc

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only