Learn More
3M™ Disposable Maintenance-Free Half Mask Respirator, 4000 Series
Offers reliable respiratory protection against a range of gases, vapors, and particulates. The 3M Disposable Maintenance-Free Half Mask Respirator can be used until damaged, particulate filters clog, gas filters become saturated or after one month.
27.39 € - 41.64 €
Products 5
Product Code Brand Cartridge Type Price Quantity & Availability  
Product Code Brand Cartridge Type Price Quantity & Availability  
12372399
View Documents
3M™
7000034735
N/A N/A N/A
12392399
View accessory products View Documents
3M™
7000088755
N/A N/A N/A
12302409
View Documents
3M™
7000088756
N/A N/A N/A
12312409
View Documents
3M™
7000088757
FFABEK1P3 R D N/A N/A N/A
11733343
View accessory products View Documents
3M™
7000088757
N/A N/A N/A
Description
Description
Simple, lightweight, well-balanced respirator offers reliable respiratory protection against a range of gases, vapors, and particulates.
  • No parts assembly or maintenance required
  • Unique bonded carbon technology allows the respirator to sit close to the face
  • Low profile design allows the respirator to fit under many other types of protective equipment, providing an undisturbed field of vision and a more comfortable worklife
  • Exhalation valve centrally positioned for effective removal of heat and moisture
  • Can be used until damaged, particulate filters clog, gas filters become saturated or after one month
Respiratory protection against:
  • Fine dusts
  • Oil and water based mists
  • Organic gases or vapours with good warning properties and boiling point above 65°C
Approved to:
  • EN 405:2002
  • Maximum usage limitations:
  • Gases/vapours: APF - 10 x WEL
  • Particulates: APF - 20 x WEL
  • CE marked 
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Disposable Maintenance-Free Half Mask Respirator, 4000 Series