Learn More
Uvex™ Disposable Polypropylene Chemical Protection Hood
Disposable chemical protection hood constructed of spunbond polypropylene laminated with polyethylene film. One size fits most, for single use only.
Brand: Uvex™ 9875200
76.38 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- spunbond polypropylene laminated with polyethylene film
- Hook and loop closure
- One size fits most
- Single use only
Specifications
Hood | |
Spunbond Polypropylene | |
Hook and Loop | |
Non-sterile |
Universal | |
White | |
Single-use |