missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Red Disposable Protective Coveralls, 4515 Series
Provides PPE Directive Category III, Type 5/6 protection. The Protective Coverall 4515 Series protect from hazardous dusts such as asbestos fibers.
72.64 € - 99.76 €
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | SMS Polypropylene |
Color | Red |
Disposable | Single-use |
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Height (English) | Height (Metric) | Price | Quantity & Availability | |||
14840982
|
3M™
7000089631 |
4X-Large | 78 to 81 in. | 200 to 206 cm | N/A | ||||
14890972
|
3M™
7000089626 |
Medium | 66 to 69 in. | 167 to 176 cm | N/A | ||||
14810982
|
3M™
7000089628 |
X-Large | 70 to 74 in. | 179 to 187 cm | N/A | ||||
14860972
|
3M™
7000089625 |
Small | 64 to 67 in. | 164 to 170 cm | N/A | ||||
14820982
|
3M™
7000089629 |
2X-Large | 73 to 76 in. | 186 to 194 cm | N/A | ||||
14830982
|
3M™
7000089630 |
3X-Large | 76 to 78 in. | 194 to 200 cm | N/A | ||||
14800982
|
3M™
7000089627 |
Large | 69 to 71 in. | 174 to 181 cm | N/A | ||||
Description
- Breathable material reduces heat build-up and discomfort
- Excellent barrier against dry particles and limited liquid splash (CE Type 5/6)
- Robust, highly breathable 50gsm SMS material
- Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
- Two-way zipper with fully sealable storm flap
- Two-panel hood design
- Elasticated wrists, waist, and ankles for increased comfort and freedom of movement
Specifications
General Purpose Coveralls | |
SMS Polypropylene | |
Single-use | |
Elastic | |
2-panel | |
Serged | |
PPE Directive Category III, Type 5/6 |
Unisex | |
Red | |
Elastic | |
Elastic | |
Front Zipper with Storm Flap | |
Non-sterile |