Learn More
Kartell™ Disposable Stirrer for Cuvets
Manufactured from polystyrene to avoid breakage of fragile cuvets. Kartell™ Disposable Stirrers for Cuvets are designed with a rounded end for comfort and a flat stirring paddle for optimum mixing.
Brand:  Kartell™ 0093900
Product Code. 10315921
  • 5.45 € / Pack of 100
Estimated Shipment: 23-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Manufactured from polystyrene to avoid breakage of fragile cuvets. Kartell™ Disposable Stirrers for Cuvets are designed with a rounded end for comfort and a flat stirring paddle for optimum mixing.
  • CE-IVD certified
  • Disposable
Specifications
Specifications
CE-IVD
90 mm
Stirrer
3 mm
Polystyrene