missing translation for 'onlineSavingsMsg'
Learn More

Simport™ Disposable Transfer Pipets

Product Code. p-8000443
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Pack of 4000
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 13496408

Brand: Simport™ P2005610S

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Eliminate the hazard of broken glass and exposure to infectious materials with these all-in-one pipets. Simport™ Disposable Transfer Pipets are molded from see-through low density polyethylene and inert to biological fluids and most acids.

  • LDPE transfer pipets
  • Put an end to matching rubber bulbs with glass pipets
  • Can be sealed and refrigerated
  • Work well whenever there is a need for quick, safe transfer of fluids
  • Temperature resistant down to -196 °C. Can be gaz (EtO) sterilized
  • Choose between 12 popular models, available in several sizes, tip designs and lengths, in sterile or non-sterile packaging
  • Available with graduations

Specifications

Bulb Draw (Metric) 1.9 mL
Diameter (Metric) Outer 6.7 mm
Material LDPE
Drops/mL 25
For Use With (Application) For quick, safe transfer of fluids
Graduated Yes
Quantity 10 Pack
Volume (Metric) 5 mL
Length (Metric) 15.5 cm
Type Transfer Pipet
Disposable Single-use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.