Learn More
Dissecting Set
Use this dissecting kit for a variety of applications. Dissecting Sets are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand: S Murray SAMCO Y261/74
This item is not returnable.
View return policy
Description
Stainless steel instruments in a wallet.
Kit includes:
- 1 Scalpel, Polished Blade, SS Light Pattern, 35mm
- 1 Scalpel, Polished Blade, SS Light Pattern, 45mm
- 2 Dissecting Needles, all stainless steel
- 1 PVC Dissecting Roll
- 1 Spring Forceps Blunt, SS, 125mm
- 1 Spring Forceps Sharp with Guide Pin, SS, 125mm
- 1 Dissecting Scissors, Open Shanks, SS, 112mm
- 1 Magnifier
Specifications
Dissecting Set | |
Stainless Steel | |
1 Scalpel (35mm), 1 Scalpel (45mm), 2 Dissecting Needles, 1 PVC Dissecting Roll, 1 Spring Forceps Blunt (125mm), 1 Spring Forceps Sharp w/ Guide Pin (125mm), 1 Dissecting Scissors (Open Shanks, 112mm), 1 Magnifier |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Dissecting Set > Stainless Steel
Spot an opportunity for improvement?Share a Content Correction