Learn More
Fisherbrand™ Dissecting Scissors
Use these scissors for a variety of applications. S Murray™ Dissecting Scissors are suitable for theatre, ward, veterinary, dental and chiropody use.
Brand:  Fisherbrand™ E107/01
Product Code. 15207266
  • 28.26 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Straight, stainless steel dissecting scissors
  • Closed shanks, fine point
  • Comply with the essential requirements of the Medical Devices Directive 93/42/EEC
  • Full traceability maintained throughout the manufacturing process.
  • Made from only high grade stainless steel in accordance to the specifications of ISO 7153-1
  • 10 year guarantee
Specifications
Specifications
Dissecting Scissors
Straight
105 mm
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Dissecting Scissors > Dissecting Scissor