missing translation for 'onlineSavingsMsg'
Learn More

Distillation Standard ASTM D 86; NF EN ISO 3405 (Nominal value: 164-357 °C), Paragon Scientific™

Product Code. 16080182
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 16080182

Brand: Paragon Scientific CRMDIGO

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Distillation Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products for Distillation according to ASTM D86. Alternative test methods are IP 123 and ISO 3405

  • Certified Reference Material for Distillation
  • Manufactured in accordance with ASTM / EN or IP test methods
  • Certified via Round Robin
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • 12 month shelf life.
  • Manufactured in the United Kingdom

Specifications

UN Number UN 1202
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.

Nominal values quoted. Certified Values will vary from batch to batch.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.