missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Distilling Links
Distilling Links for the connection of distillation thermometers of 55 mm built-in length.
Brand: Lenz Laborglasinstrumente™ 05053314
This item is not returnable.
View return policy
Description
- With Liebig condenser
- Outlet with NS cone
- Water connection with glass olives, with NS socket
Specifications
For distillation thermometers of 55 mm built-In length | |
Duran Glass | |
250 mm |
Distilling Link | |
NS 29/32, GL 14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Distilling Links > Length: 250 mm; NS 29/32, GL 14
Spot an opportunity for improvement?Share a Content Correction