Learn More
Lenz Laborglasintrumente Distilling Links with Splash Head
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
124.66 € - 158.26 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10244341
|
Lenz Laborglasintrumente
05043001 |
150 mm |
124.66 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10020231
|
Lenz Laborglasintrumente
05043003 |
300 mm |
158.26 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10335931
|
Lenz Laborglasintrumente
05043002 |
200 mm |
134.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- With two vertical NS cones and splash head
- Made of DURAN™ tubing
Specifications
Borosilicate Glass 3.3 |