missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasintrumente Distilling Receivers Acc. to Anschütz Thiele, Straight, with Cooling Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
781.22 € - 800.09 €
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Capacity (Metric) | Price | Quantity & Availability | ||||
10683622
|
Lenz Laborglasintrumente
05305014 |
Duran Glass | 100 mL |
789.59 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10000491
|
Lenz Laborglasintrumente
05305004 |
Duran Glass | 50 mL |
781.22 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10244531
|
Lenz Laborglasintrumente
05305104 |
Duran Tubing, PTFE | 50 mL |
793.62 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10468212
|
Lenz Laborglasintrumente
05305114 |
Duran Tubing, PTFE | 100 mL |
800.09 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Receivers with straight adapter
- With cooling jacket, graduated, with socket and cone NS 29/32
- Vacuum connection: thread GL18
- NS stopcock with solid glass or PTFE plug
- Made of DURAN™ tubing
Specifications
Socket and Cone NS 29/32 |