missing translation for 'onlineSavingsMsg'
Learn More
Disuccinimidyl glutarate, 97%, Thermo Scientific Chemicals Product Code.: 15404538

Disuccinimidyl glutarate, 97%, Thermo Scientific Chemicals

Product Code. 15404538
100 mg
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy

Product Code. 15404538

Brand: Thermo Scientific Alfa Aesar H58208.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Disuccinimidyl glutarate is used as a membrane-soluble, amine-reactive N-hydroxysuccinamide ester-based homobifunctional crosslinking reagent. It is a homobifunctionla cross-linking reagent, for ex. isolation of the insulin receptor.

Solubility
Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79642-50-5
Molecular Formula C13H14N2O8
Molecular Weight (g/mol) 326.26
MDL Number MFCD00153597
InChI Key LNQHREYHFRFJAU-UHFFFAOYSA-N
Synonym disuccinimidyl glutarate, di n-succinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl pentanedioate, pentanedioic acid,1,5-bis 2,5-dioxo-1-pyrrolidinyl ester, dsg crosslinker, glutaric acid disuccinimidyl ester, di n-hydroxysuccinimidyl glutarate, bis 2,5-dioxopyrrolidin-1-yl glutarate, di n-succinimidyl glutarate chn
PubChem CID 4432628
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) pentanedioate
SMILES O=C(CCCC(=O)ON1C(=O)CCC1=O)ON1C(=O)CCC1=O

Specifications

Quantity 100 mg
Beilstein 8575145
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethylsulfoxide and dimethylformamide. Partially soluble in water.
Formula Weight 326.26
Percent Purity 97%
Chemical Name or Material Disuccinimidyl glutarate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP11L039 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog NumberH58208.ME,H58208.MC
Lot NumberM23L014 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberH58208.ME,H58208.MC
Lot NumberZ10K038 Certificate TypeCertificate of Analysis Date11/12/2024 Catalog NumberH58208.ME,H58208.MC
Lot NumberT25K034 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog NumberH58208.ME,H58208.MC
Lot NumberQ29K040 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog NumberH58208.ME,H58208.MC
compliance-icons
Product Identifier
  • Disuccinimidyl glutarate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Disuccinimidyl glutarate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Disuccinimidyl glutarate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.