Learn More
Tocris Bioscience™ Disulfide biotin azide
Azide-activated biotinylation reagent; labels alkyne moieties via copper-catalyzed click chemistry
Brand:  Tocris Bioscience™ 7768/5
Product Code. 18728296
  • 432.00 € / 5mg
Explore more special offers
Description
Description
Disulfide Biotin Azide is an azide-activated cleavable (disulphide) biotin probe. It allows the biotinylation of molecules containing alkyne moiety via copper-catalyzed click chemistry reactions.
Specifications
Specifications
N-(24-Azido-12,20-dioxo-3,6,9-trioxa-16,17-dithia-13,21-diazatetracosyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide
5 mg
C27H48N8O7S3
2866429-93-6
General Laboratory Research Reagent