missing translation for 'onlineSavingsMsg'
Learn More
Dithiooxamide, 98%, Thermo Scientific Chemicals Product Code.: 15700799

Dithiooxamide, 98%, Thermo Scientific Chemicals

Product Code. 15700799
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15700799

Brand: Thermo Scientific Acros 117551000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-40-3
Molecular Formula C2H4N2S2
Molecular Weight (g/mol) 120.19
InChI Key OAEGRYMCJYIXQT-UHFFFAOYSA-N
Synonym dithiooxamide, rubeanic acid, ethanebis thioamide, oxamide, dithio, dithioxamide, rubeane, rubean, hydrorubeanic acid, oxaldiimidic acid, dithio, usaf mk-6Show MoreShow Less
PubChem CID 2777982
IUPAC Name ethanedithioamide
SMILES C(=S)(C(=S)N)N

Specifications

Melting Point 200.0°C
Elemental Analysis 22.7 to 23.9% (N) 19.5 to 20.5% (C)
Infrared Spectrum Authentic
Linear Formula H2NCSCSNH2
Quantity 100 g
Merck Index 15,8412
Solubility Information Solubility in water: 0.37g/L (20°C).
Formula Weight 120.19
Percent Purity 98%
Chemical Name or Material Dithiooxamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467534 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number117550050,117551000,117550250
Lot NumberA0453934 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number117550050,117551000,117550250
Lot NumberA0457456 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number117550050,117551000,117550250
Lot NumberA0453934 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number117550050,117551000,117550250
Lot NumberA0432287 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number117550050,117551000,117550250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dithiooxamide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.